MAN182108640 Manchester Organics

4-Chloro-3-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD06660219
  • Molecular Weight : 224.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P18486

Safety Information

MSDS https://www.manchesterorganics.com/P18486
Availability details for
Quantity
  • Availability
  • Add to Cart