MAN182108645 Manchester Organics

(2,6-Dimethyl-pyridin-4-yl)-methanol

  • MDL number MFCD00956275
  • Molecular Weight : 137.17903

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K18269

Safety Information

MSDS https://www.manchesterorganics.com/K18269
Availability details for
Quantity
  • Availability
  • Add to Cart