MAN182108661 Manchester Organics

Methyl 2-amino-3-chlorobenzoate

  • MDL number MFCD06797833
  • Cas Number : 77820-58-7
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/X18328
Availability details for
Quantity
  • Availability
  • Add to Cart