MAN182108664 Manchester Organics

2-Amino-5,6-dihydro-4-[4H]-cyclopenta[B]thiophene-3-carboxylic acid ethyl ester

  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Inchi Code 1S/C10H13NO2S/c1-2-13-10(12)8-6-4-3-5-7(6)14-9(8)11/h2-5,11H2,1H3
Inchi Key BOJXCJDYZJSPMZ-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/A18331
Availability details for
Quantity
  • Availability
  • Add to Cart