MAN182108716 Manchester Organics

2-Chloro-3-fluorobenzoic acid

  • MDL number MFCD00973903
  • Molecular Weight : 174.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E18429

Safety Information

MSDS https://www.manchesterorganics.com/E18429
Availability details for
Quantity
  • Availability
  • Add to Cart