MAN182108729 Manchester Organics

2-Methylpiperazine-1-carboxylic acid benzyl ester

  • MDL number MFCD04115058
  • Molecular Weight : 234.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V18704

Safety Information

MSDS https://www.manchesterorganics.com/V18704
Availability details for
Quantity
  • Availability
  • Add to Cart