MAN182108735 Manchester Organics

N-1-Benzyl-3-methylpiperazine

  • MDL number MFCD01570796
  • Cas Number : 3138-90-7
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-3-methylpiperazine
Inchi Code 1S/C12H18N2/c1-11-9-14(8-7-13-11)10-12-5-3-2-4-6-12/h2-6,11,13H,7-10H2,1H3
Inchi Key QOFUDSPYJDXBOF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y18375
Availability details for
Quantity
  • Availability
  • Add to Cart