MAN182108753 Manchester Organics

N-1-Boc-2-phenylpiperazine

  • MDL number MFCD07367734
  • Molecular Weight : 262.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C18427

Safety Information

MSDS https://www.manchesterorganics.com/C18427
Availability details for
Quantity
  • Availability
  • Add to Cart