MAN182108754 Manchester Organics

N-1-Boc-2-benzylpiperazine

  • MDL number MFCD06739084
  • Cas Number : 481038-63-5
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D18382
Availability details for
Quantity
  • Availability
  • Add to Cart