MAN182108788 Manchester Organics

2-Fluoro-6-(trifluoromethyl)pyridine

  • MDL number MFCD03092901
  • Cas Number : 94239-04-0
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K18411
Availability details for
Quantity
  • Availability
  • Add to Cart