MAN182108807 Manchester Organics

6-Methyl-2,3,5-tribromopyridine

  • MDL number MFCD01646071
  • Cas Number : 3430-15-7
  • Molecular Weight : 329.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W18421
Availability details for
Quantity
  • Availability
  • Add to Cart