MAN182108823 Manchester Organics

N-Ethyl-7-nitro-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD07368631
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C18475

Safety Information

MSDS https://www.manchesterorganics.com/C18475
Availability details for
Quantity
  • Availability
  • Add to Cart