MAN182108832 Manchester Organics

4-Amino-5-hydroxymethyl-2-(methylthio)pyrimidine

  • MDL number MFCD08236793
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K18505

Safety Information

MSDS https://www.manchesterorganics.com/K18505
Availability details for
Quantity
  • Availability
  • Add to Cart