MAN182108837 Manchester Organics

4-n-Propylpyridine

  • MDL number MFCD00014639
  • Cas Number : 1122-81-2
  • Molecular Weight : 121.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/S18440
Availability details for
Quantity
  • Availability
  • Add to Cart