MAN182108842 Manchester Organics

4-Diethylamino-2-methoxybenzaldehyde

  • MDL number MFCD02277405
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F18288
Availability details for
Quantity
  • Availability
  • Add to Cart