MAN182108867 Manchester Organics

4-Bromo-2-(difluoromethyl)-1-fluorobenzene

  • MDL number MFCD06411633
  • Cas Number : 445303-69-5
  • Molecular Weight : 225.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K18459
Availability details for
Quantity
  • Availability
  • Add to Cart