MAN182108870 Manchester Organics

4-Benzyloxy-2-methoxybenzaldehyde

  • MDL number MFCD01075699
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P18463

Safety Information

MSDS https://www.manchesterorganics.com/P18463
Availability details for
Quantity
  • Availability
  • Add to Cart