MAN182108871 Manchester Organics

(4-Chloro-3-nitrophenyl)-(4-fluorophenyl)-methanone

  • MDL number MFCD03840494
  • Molecular Weight : 279.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R18464

Safety Information

MSDS https://www.manchesterorganics.com/R18464
Availability details for
Quantity
  • Availability
  • Add to Cart