MAN182108872 Manchester Organics

5-Bromopicolinic acid

  • MDL number MFCD00234149
  • Cas Number : 30766-11-1
  • Molecular Weight : 202.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/L16021
Availability details for
Quantity
  • Availability
  • Add to Cart