MAN182108875 Manchester Organics

6-Hydroxypyrimidine-4-carboxylic acid

  • MDL number MFCD00023267
  • Cas Number : 6299-87-2
  • Molecular Weight : 140.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-oxo-1,6-dihydro-4-pyrimidinecarboxylic acid
Inchi Code 1S/C5H4N2O3/c8-4-1-3(5(9)10)6-2-7-4/h1-2H,(H,9,10)(H,6,7,8)
Inchi Key QYGHXDAYBIFGKI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T18299
Availability details for
Quantity
  • Availability
  • Add to Cart