MAN182108881 Manchester Organics

2-Chloro-1-N-isopropylimidazole

  • MDL number MFCD10699127
  • Molecular Weight : 144.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z18802

Safety Information

MSDS https://www.manchesterorganics.com/Z18802
Availability details for
Quantity
  • Availability
  • Add to Cart