MAN182108908 Manchester Organics

2-Chloro-3,5-dibromopyridine

  • MDL number MFCD00233991
  • Cas Number : 40360-47-2
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dibromo-2-chloropyridine
Inchi Code 1S/C5H2Br2ClN/c6-3-1-4(7)5(8)9-2-3/h1-2H
Inchi Key PYSICVOJSJMFKP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/A18306
Availability details for
Quantity
  • Availability
  • Add to Cart