MAN182108910 Manchester Organics

4-Fluoro-3-methoxyphenylacetic acid

  • MDL number MFCD08146650
  • Cas Number : 946713-86-6
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-fluoro-3-methoxyphenyl)acetic acid
Inchi Code 1S/C9H9FO3/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4H,5H2,1H3,(H,11,12)
Inchi Key YNNVYROHVVBVFI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E18312
Availability details for
Quantity
  • Availability
  • Add to Cart