MAN182108922 Manchester Organics

3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole

  • MDL number MFCD03066187
  • Molecular Weight : 197.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A10924

Safety Information

MSDS https://www.manchesterorganics.com/A10924
Availability details for
Quantity
  • Availability
  • Add to Cart