MAN182108924 Manchester Organics

1-Methyl-1,2,4-triazole

  • MDL number MFCD01076192
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G18550

Safety Information

MSDS https://www.manchesterorganics.com/G18550
Availability details for
Quantity
  • Availability
  • Add to Cart