MAN182108960 Manchester Organics

1-Ethyl-2-chloro-1N-imidazole

  • MDL number MFCD10699124
  • Molecular Weight : 130.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L18602

Safety Information

MSDS https://www.manchesterorganics.com/L18602
Availability details for
Quantity
  • Availability
  • Add to Cart