MAN182108981 Manchester Organics

4-Phenoxypiperidine

  • MDL number MFCD04114971
  • Molecular Weight : 177.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U18822

Safety Information

MSDS https://www.manchesterorganics.com/U18822
Availability details for
Quantity
  • Availability
  • Add to Cart