MAN182108986 Manchester Organics

1H-Benzimidazole-2-sulfonic acid

  • MDL number MFCD00100972
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z17762

Safety Information

MSDS https://www.manchesterorganics.com/Z17762
Availability details for
Quantity
  • Availability
  • Add to Cart