MAN182108997 Manchester Organics

2-Amino-4-methylpyrimidine

  • MDL number MFCD00006101
  • Cas Number : 108-52-1
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C18523
Availability details for
Quantity
  • Availability
  • Add to Cart