MAN182109013 Manchester Organics

7-Benzyloxy-4-chloro-6-methoxy-quinazoline

  • MDL number MFCD04115120
  • Molecular Weight : 300.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R18700

Safety Information

MSDS https://www.manchesterorganics.com/R18700
Availability details for
Quantity
  • Availability
  • Add to Cart