MAN182109016 Manchester Organics

2,4-Difluoro-3-methylbenzaldehyde

  • MDL number MFCD07777152
  • Molecular Weight : 156.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R16760

Safety Information

MSDS https://www.manchesterorganics.com/R16760
Availability details for
Quantity
  • Availability
  • Add to Cart