MAN182109029 Manchester Organics

4-Isopropylbenzenesulfonamide

  • MDL number MFCD00457071
  • Cas Number : 6335-39-3
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B10925
Availability details for
Quantity
  • Availability
  • Add to Cart