MAN182109041 Manchester Organics

2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile

  • MDL number MFCD08445711
  • Molecular Weight : 223.6556

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W10659

Safety Information

MSDS https://www.manchesterorganics.com/W10659
Availability details for
Quantity
  • Availability
  • Add to Cart