MAN182109055 Manchester Organics

3-Fluoro-5-methylphenylacetonitrile

  • MDL number MFCD03094317
  • Molecular Weight : 149.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P18722

Safety Information

MSDS https://www.manchesterorganics.com/P18722
Availability details for
Quantity
  • Availability
  • Add to Cart