MAN182109056 Manchester Organics

3,5-Bis(trifluoromethyl)anisole

  • MDL number MFCD00728736
  • Molecular Weight : 244.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R18723

Safety Information

MSDS https://www.manchesterorganics.com/R18723
Availability details for
Quantity
  • Availability
  • Add to Cart