MAN182109069 Manchester Organics

(2,6-Dichloropyrimidin-4-yl)-(4-fluorophenyl)methanone

  • MDL number MFCD11100517
  • Cas Number : 1099597-81-5
  • Molecular Weight : 271.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-dichloro-4-pyrimidinyl)(4-fluorophenyl)methanone
Inchi Code 1S/C11H5Cl2FN2O/c12-9-5-8(15-11(13)16-9)10(17)6-1-3-7(14)4-2-6/h1-5H
Inchi Key QIULZDCQGAXJIB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/V10917
Availability details for
Quantity
  • Availability
  • Add to Cart