MAN182109101 Manchester Organics

3,5-Bis(trifluoromethyl)phenylacetic acid ethyl ester

  • MDL number MFCD08689894
  • Cas Number : 144632-97-3
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl [3,5-bis(trifluoromethyl)phenyl]acetate
Inchi Code 1S/C12H10F6O2/c1-2-20-10(19)5-7-3-8(11(13,14)15)6-9(4-7)12(16,17)18/h3-4,6H,2,5H2,1H3
Inchi Key ICTVPOAERFKXSS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/F10526
Availability details for
Quantity
  • Availability
  • Add to Cart