MAN182109151 Manchester Organics

2-[4-Fluoro-3-(trifluoromethyl)phenyl]propan-2-ol

  • MDL number MFCD11226509
  • Cas Number : 534-39-4
  • Molecular Weight : 222.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[4-fluoro-3-(trifluoromethyl)phenyl]-2-propanol
Inchi Code 1S/C10H10F4O/c1-9(2,15)6-3-4-8(11)7(5-6)10(12,13)14/h3-5,15H,1-2H3
Inchi Key QQNITPNLHMKSLF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/B10923
Availability details for
Quantity
  • Availability
  • Add to Cart