MAN182109161 Manchester Organics

1-Bromo-3-(2,2-difluoropropyl)benzene

  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-(2,2-difluoropropyl)benzene
Inchi Code 1S/C9H9BrF2/c1-9(11,12)6-7-3-2-4-8(10)5-7/h2-5H,6H2,1H3
Inchi Key SAHNEOQOFLAPRM-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/R15102
Availability details for
Quantity
  • Availability
  • Add to Cart