MAN182109162 Manchester Organics

1-Bromo-4-(2,2-difluoropropyl)benzene

  • MDL number MFCD11226517
  • Cas Number : 1099598-20-5
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-(2,2-difluoropropyl)benzene
Inchi Code 1S/C9H9BrF2/c1-9(11,12)6-7-2-4-8(10)5-3-7/h2-5H,6H2,1H3
Inchi Key VIEJQUIQUJATPR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S15103
Availability details for
Quantity
  • Availability
  • Add to Cart