MAN182109167 Manchester Organics

4-Chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD11226523
  • Cas Number : 1099598-24-9
  • Molecular Weight : 212.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-1-fluoro-2-(2,2,2-trifluoroethyl)benzene
Inchi Code 1S/C8H5ClF4/c9-6-1-2-7(10)5(3-6)4-8(11,12)13/h1-3H,4H2
Inchi Key HIIQQIYUDSBFOB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z15110
Availability details for
Quantity
  • Availability
  • Add to Cart