MAN182109177 Manchester Organics

1-Fluoro-4-isopropenyl-2-(trifluoromethyl)benzene

  • MDL number MFCD11226534
  • Molecular Weight : 204.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A10924
Availability details for
Quantity
  • Availability
  • Add to Cart