MAN182109184 Manchester Organics

2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride

  • MDL number MFCD10565658
  • Molecular Weight : 229.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P15291

Safety Information

MSDS https://www.manchesterorganics.com/P15291
Availability details for
Quantity
  • Availability
  • Add to Cart