MAN182109189 Manchester Organics

1-(2,2,2-Trifluoroethyl)-4-trifluoromethylbenzene

  • MDL number MFCD11226545
  • Cas Number : 157928-43-3
  • Molecular Weight : 228.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A15136
Availability details for
Quantity
  • Availability
  • Add to Cart