MAN182109190 Manchester Organics

3-Chloro-2,6-difluorobenzotrifluoride

  • MDL number MFCD11226546
  • Cas Number : 1099597-36-0
  • Molecular Weight : 216.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-chloro-2,4-difluoro-3-(trifluoromethyl)benzene
Inchi Code 1S/C7H2ClF5/c8-3-1-2-4(9)5(6(3)10)7(11,12)13/h1-2H
Inchi Key ZABHDJHDUYGAPB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/U15130
Availability details for
Quantity
  • Availability
  • Add to Cart