MAN182109203 Manchester Organics

5,6,7,8-Tetrahydro-quinolin-8-ol

  • MDL number MFCD07369987
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T18631
Availability details for
Quantity
  • Availability
  • Add to Cart