MAN182109221 Manchester Organics

5-Bromo-2-chloropyridine-4-carboxylic acid

  • MDL number MFCD07375114
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V18656
Availability details for
Quantity
  • Availability
  • Add to Cart