MAN182109224 Manchester Organics

4,6-Difluoroindole

  • MDL number MFCD01075213
  • Molecular Weight : 153.1288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X18658

Safety Information

MSDS https://www.manchesterorganics.com/X18658
Availability details for
Quantity
  • Availability
  • Add to Cart