MAN182109254 Manchester Organics

5-Bromo-2,4-bis(trifluoromethyl)pyridine

  • MDL number MFCD11100546
  • Cas Number : 1099598-05-6
  • Molecular Weight : 293.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U15034
Availability details for
Quantity
  • Availability
  • Add to Cart