MAN182109277 Manchester Organics

3-Fluoro-5-(trifluoromethyl)pyridine

  • MDL number MFCD11100535
  • Cas Number : 1099597-96-2
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/N15029
Availability details for
Quantity
  • Availability
  • Add to Cart